Stampa…

4-Thiazolecarbonyl chloride 95%

Fornitore: Apollo Scientific
Pericolo

OR6374-1G OR6374-100MG
APOSOR6374-1GEA 198 EUR
APOSOR6374-1G APOSOR6374-100MG
4-Thiazolecarbonyl chloride 95%
4-Thiazolecarbonyl chloride
Formula: C₄H₂ClNOS
Temperatura di stoccaggio: Refrigerata
CAS Number: 3745-79-7
UN: 3261
ADR: 8,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR