1-(4-Chlorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Fornitore: Apollo Scientific
OR6329-1G
OR6329-5G
OR6329-25G
APOSOR6329-1GEA
43.5
EUR
APOSOR6329-1G
APOSOR6329-5G
APOSOR6329-25G
1-(4-Chlorophenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(4-Chlorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Formula:
C₁₁H₁₀ClNO₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00158023 CAS Number: 39629-87-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...