(4-Methylpyridin-2-yl)methylamine 95%
Fornitore: Apollo Scientific
|
Pericolo
|
Formula:
C₇H₁₀N₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD06213857 CAS Number: 129768-95-2 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...