Stampa…

1-(3-Bromophenyl)cyclopropanecarboxylic acid

Fornitore: Apollo Scientific

OR61221-1G OR61221-250MG
APOSOR61221-1GEA 69.8 EUR
APOSOR61221-1G APOSOR61221-250MG
1-(3-Bromophenyl)cyclopropanecarboxylic acid
1-(3-Bromophenyl)cyclopropanecarboxylic acid
Formula: C₁₀H₉BrO₂
Peso molecolare: 241,0813 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07374441
CAS Number: 124276-95-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR