3'-Bromo-4'-morpholinoacetophenone
Fornitore: Apollo Scientific
OR59418-1G
OR59418-250MG
APOSOR59418-1GEA
161
EUR
APOSOR59418-1G
APOSOR59418-250MG
3'-Bromo-4'-morpholinoacetophenone
3'-Bromo-4'-morpholinoacetophenone
Formula:
C₁₂H₁₄BrNO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09878364 CAS Number: 957066-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...