4-Bromo-N-cyclopropylbenzenesulphonamide 98%
Fornitore: Apollo Scientific
Formula:
C₉H₁₀BrNO₂S Punto di fusione: 110…112 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00463486 CAS Number: 331950-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...