N,N,N',N',N''-Pentamethyldiethylenetriamine
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine
OR55651-100G
OR55651-500G
APOSOR55651-100GEA
21.9
EUR
APOSOR55651-100G
APOSOR55651-500G
N,N,N',N',N''-Pentamethyldiethylenetriamine
N,N,N',N',N''-Pentamethyldiethylenetriamine
Formula:
C₉H₂₃N₃ Peso molecolare: 173,30 g/mol Punto di ebollizione: 198 °C Punto di fusione: –20 °C Densità: 0,83 g/cm³ (25 °C) Flash Pt: 77 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00014876 CAS Number: 3030-47-5 UN: 2922 ADR: 8+6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...