Stampa…

4-Ethynyltetrahydro-2H-pyran 97%

Fornitore: Apollo Scientific
Pericolo

OR55577-1G OR55577-250MG OR55577-5G
APOSOR55577-1GEA 221 EUR
APOSOR55577-1G APOSOR55577-250MG APOSOR55577-5G
4-Ethynyltetrahydro-2H-pyran 97%
4-Ethynyltetrahydro-2H-pyran
Formula: C₇H₁₀O
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD17016081
CAS Number: 1202245-65-5
UN: 1993
ADR: 3,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR