Stampa…

3-Propoxybenzoic acid 98%

Fornitore: Apollo Scientific

OR55479-1G OR55479-5G
APOSOR55479-1GEA 168 EUR
APOSOR55479-1G APOSOR55479-5G
3-Propoxybenzoic acid 98%
3-Propoxybenzoic acid
Formula: C₁₀H₁₂O₃
Temperatura di stoccaggio: Ambiente
CAS Number: 190965-42-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR