4-(4-Bromophenyl)dibenzofuran
Fornitore: Apollo Scientific
Synonyms:
4-(4-Bromophenyl)dibenzo[b,d]furan
OR55286-1G
OR55286-5G
APOSOR55286-1GEA
66.9
EUR
APOSOR55286-1G
APOSOR55286-5G
4-(4-Bromophenyl)dibenzofuran
4-(4-Bromophenyl)dibenzofuran
Formula:
C₁₈H₁₁BrO Peso molecolare: 323,19 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD19441306 CAS Number: 955959-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...