Stampa…

Octenidine dihydrochloride 98%

Fornitore: Apollo Scientific

Synonyms: 1,1'-(Decane-1,10-diyl)bis(N-octylpyridin-4(1H)-imine) dihydrochloride

OR54413-100G OR54413-25G OR54413-5G
APOSOR54413-100GEA 720 EUR
APOSOR54413-100G APOSOR54413-25G APOSOR54413-5G
Octenidine dihydrochloride 98%
Octenidine dihydrochloride
Formula: C₃₆H₆₂N₄
Peso molecolare: 623,84 g/mol
Punto di fusione: 214 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01938808
CAS Number: 70775-75-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR