3-(Benzyloxy)pyrrolidine
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₁₁H₁₅NO Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD08060975 CAS Number: 420137-14-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...