Malonamide 98%
Fornitore: Apollo Scientific
|
Attenzione
|
OR5304-100G
OR5304-500G
APOSOR5304-100GEA
114
EUR
APOSOR5304-100G
APOSOR5304-500G
Malonamide 98%
Malonamide
Reagent for fluorimetric determination of reducing carbohydrates: Anal. Chim. Acta, 108, 421 (1979).
Formula:
C₃H₆N₂O₂ Peso molecolare: 102,09 g/mol Punto di fusione: 172…175 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008034 CAS Number: 108-13-4 EINECS: 203-553-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...