Stampa…

4-Bromo-2-piperazinopyridine 96%

Fornitore: Apollo Scientific

Synonyms: 1-(4-Bromo-2-pyridyl)piperazine

OR52759-1G OR52759-5G
APOSOR52759-1GEA 219 EUR
APOSOR52759-1G APOSOR52759-5G
4-Bromo-2-piperazinopyridine 96%
4-Bromo-2-piperazinopyridine
Formula: C₉H₁₂BrN₃
Peso molecolare: 242,1157 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD16618975
CAS Number: 1201643-59-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR