5-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine
Fornitore: Apollo Scientific
|
Pericolo
|
OR52607-1G
OR52607-5G
APOSOR52607-1GEA
64
EUR
APOSOR52607-1G
APOSOR52607-5G
5-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine
5-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine
Formula:
C₆H₅BrN₄ Peso molecolare: 213,0400 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11655928 CAS Number: 1010120-55-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...