2-(Benzyloxy)-1-ethanamine 96%
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
2-(Benzyloxy)ethanamine, 2-(Benzyloxy)ethylamine
Formula:
C₉H₁₃NO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00235180 CAS Number: 38336-04-8 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...