Stampa…

N-Methoxycarbonylmaleimide

Fornitore: Apollo Scientific
Attenzione

Synonyms: Methyl 2,5-dioxo-2,5-dihydro-1H-pyrrole-1-carboxylate

OR51792-1G OR51792-5G
APOSOR51792-1GEA 59.7 EUR
APOSOR51792-1G APOSOR51792-5G
N-Methoxycarbonylmaleimide
N-Methoxycarbonylmaleimide
Formula: C₆H₅NO₄
Peso molecolare: 155,11 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00042755
CAS Number: 55750-48-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR