Stampa…

3-Chloro-p-toluic acid 97%

Fornitore: Apollo Scientific

OR5157-100G OR5157-25G OR5157-10G
APOSOR5157-100GEA 87 EUR
APOSOR5157-100G APOSOR5157-25G APOSOR5157-10G
3-Chloro-p-toluic acid 97%
3-Chloro-p-toluic acid
Formula: C₈H₇ClO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00045841
CAS Number: 5162-82-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR