Stampa…

N,N'-Di(1-naphthyl)benzidine 99%

Fornitore: Apollo Scientific

Synonyms: N,N’-Di(1-naphthyl)-4,4’-benzidine

OR51019-1G OR51019-5G
APOSOR51019-1GEA 36.4 EUR
APOSOR51019-1G APOSOR51019-5G
N,N'-Di(1-naphthyl)benzidine 99%
N,N'-Di(1-naphthyl)benzidine
Formula: C₃₂H₂₄N₂
Peso molecolare: 436,558 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09261380
CAS Number: 152670-41-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR