7-Methyl-8-nitroquinoline
Fornitore: Apollo Scientific
|
Attenzione
|
OR50941-25G
OR50941-5G
APOSOR50941-25GEA
299
EUR
APOSOR50941-25G
APOSOR50941-5G
7-Methyl-8-nitroquinoline
7-Methyl-8-nitroquinoline
Formula:
C₁₀H₈N₂O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00041234 CAS Number: 7471-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...