2-Thiopheneacetamide
Fornitore: Apollo Scientific
Synonyms:
Thiophene-2-acetamide
OR49054-100G
OR49054-25G
APOSOR49054-100GEA
292
EUR
APOSOR49054-100G
APOSOR49054-25G
2-Thiopheneacetamide
2-Thiopheneacetamide
Formula:
C₆H₇NOS Peso molecolare: 141,19 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00017556 CAS Number: 4461-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...