Stampa…

2-Bromo-5-methylpyrazine 95%

Fornitore: Apollo Scientific

OR480839-1G OR480839-5G
APOSOR480839-1GEA 161 EUR
APOSOR480839-1G APOSOR480839-5G
2-Bromo-5-methylpyrazine 95%
2-Bromo-5-methylpyrazine
Formula: C₅H₅BrN₂
Peso molecolare: 173,01 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08705754
CAS Number: 98006-90-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR