Stampa…

1-Bromo-2-naphthoic acid 98%

Fornitore: Apollo Scientific

Synonyms: 1-Bromonaphthalene-2-carboxylic acid

OR480725-1G OR480725-5G
APOSOR480725-1GEA 227 EUR
APOSOR480725-1G APOSOR480725-5G
1-Bromo-2-naphthoic acid 98%
1-Bromo-2-naphthoic acid
Formula: C₁₁H₇BrO₂
Peso molecolare: 251,0800 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00021408
CAS Number: 20717-79-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR