Stampa…

Tetrachlorocyclopropene 98%

Fornitore: Apollo Scientific

OR480662-25G OR480662-5G
APOSOR480662-25GEA 446 EUR
APOSOR480662-25G APOSOR480662-5G
Tetrachlorocyclopropene 98%
Tetrachlorocyclopropene
Formula: C₃Cl₄
Temperatura di stoccaggio: Ambiente
CAS Number: 6262-42-6

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR