Stampa…

N-Ethyl-1-naphthylamine

Fornitore: Apollo Scientific

OR480574-25G OR480574-1G OR480574-5G
APOSOR480574-25GEA 227 EUR
APOSOR480574-25G APOSOR480574-1G APOSOR480574-5G
N-Ethyl-1-naphthylamine
N-Ethyl-1-naphthylamine
Formula: C₁₂H₁₃N
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003880
CAS Number: 118-44-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR