Stampa…

2-n-Propylpiperidine 97%

Fornitore: Apollo Scientific

OR480200-1G OR480200-2G OR480200-5G
APOSOR480200-1GEA 176 EUR
APOSOR480200-1G APOSOR480200-2G APOSOR480200-5G
2-n-Propylpiperidine 97%
2-n-Propylpiperidine
Formula: C₈H₁₇N
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00056022
CAS Number: 3238-60-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR