Stampa…

Methyl-3-bromo-1H-indazole-5-carboxylate 95%

Fornitore: Apollo Scientific

Synonyms: Methyl 3-bromo-1H-indazole-5-carboxylate

OR471693-1G OR471693-5G
APOSOR471693-1GEA 78.6 EUR
APOSOR471693-1G APOSOR471693-5G
Methyl-3-bromo-1H-indazole-5-carboxylate 95%
Methyl-3-bromo-1H-indazole-5-carboxylate
Formula: C₉H₇BrN₂O₂
Peso molecolare: 255,0681 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11045401
CAS Number: 1086391-06-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR