Stampa…

(R)-(+)-2-Bromopropionic acid 98%

Fornitore: Apollo Scientific
Pericolo

Synonyms: (R)-(+)-2-Bromopropanoic acid

OR471636-10G OR471636-5G OR471636-25G OR471636-100G
APOSOR471636-10GEA 0 EUR
APOSOR471636-10G APOSOR471636-5G APOSOR471636-25G APOSOR471636-100G
(R)-(+)-2-Bromopropionic acid 98%
(R)-(+)-2-Bromopropionic acid
Formula: H₃CBrCHCOOH
Peso molecolare: 152,98 g/mol
Densità: 1,692 g/cm³ (25 °C)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00145205
CAS Number: 10009-70-8
UN: 3265
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR