2-Methyl-6-[bis(tert-butoxycarbonyl)amino]-4-chloropyrimidine
Fornitore: Apollo Scientific
Synonyms:
6-[Bis(Boc)amino]-4-chloro-2-methylpyrimidine
OR471508-1G
OR471508-5G
APOSOR471508-1GEA
256
EUR
APOSOR471508-1G
APOSOR471508-5G
2-Methyl-6-[bis(tert-butoxycarbonyl)amino]-4-chloropyrimidine
2-Methyl-6-[bis(tert-butoxycarbonyl)amino]-4-chloropyrimidine
Formula:
C₁₅H₂₂ClN₃O₄ Peso molecolare: 343,81 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD22199258 CAS Number: 1350918-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...