Stampa…

2-(6-Bromopyridin-2-yl)ethanol 95%

Fornitore: Apollo Scientific

Synonyms: 2-Bromo-6-(2-hydroxyethyl)pyridine

OR471389-1G OR471389-250MG
APOSOR471389-1GEA 273 EUR
APOSOR471389-1G APOSOR471389-250MG
2-(6-Bromopyridin-2-yl)ethanol 95%
2-(6-Bromopyridin-2-yl)ethanol
Formula: C₇H₈BrNO
Peso molecolare: 202.06 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10000073
CAS Number: 955370-07-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR