1-(Dimethylcarbamoyl)piperidine-2-carboxylic acid
Fornitore: Apollo Scientific
OR470212-1G
OR470212-5G
APOSOR470212-1GEA
194
EUR
APOSOR470212-1G
APOSOR470212-5G
1-(Dimethylcarbamoyl)piperidine-2-carboxylic acid
1-(Dimethylcarbamoyl)piperidine-2-carboxylic acid
Formula:
C₉H₁₆N₂O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD14596111 CAS Number: 1249531-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...