Stampa…

4-(4-Oxocyclohexyl)benzoic acid

Fornitore: Apollo Scientific

OR46649-1G OR46649-50MG OR46649-250MG OR46649-500MG
APOSOR46649-1GEA 459 EUR
APOSOR46649-1G APOSOR46649-50MG APOSOR46649-250MG APOSOR46649-500MG
4-(4-Oxocyclohexyl)benzoic acid
4-(4-Oxocyclohexyl)benzoic acid
Formula: C₁₃H₁₄O₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD13195602
CAS Number: 137465-01-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR