Stampa…

6-(Cyclohexyloxy)pyridin-3-amine

Fornitore: Apollo Scientific

OR46623-1G OR46623-250MG OR46623-100MG OR46623-500MG OR46623-2.5G
APOSOR46623-1GEA 345 EUR
APOSOR46623-1G APOSOR46623-250MG APOSOR46623-100MG APOSOR46623-500MG APOSOR46623-2.5G
6-(Cyclohexyloxy)pyridin-3-amine
6-(Cyclohexyloxy)pyridin-3-amine
Formula: C₁₁H₁₆N₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09729052
CAS Number: 224187-22-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR