Stampa…

3-Chloro-5-iodopyridine

Fornitore: Apollo Scientific

OR46150-100MG OR46150-1G OR46150-250MG
APOSOR46150-100MGEA 147 EUR
APOSOR46150-100MG APOSOR46150-1G APOSOR46150-250MG
3-Chloro-5-iodopyridine
3-Chloro-5-iodopyridine
Formula: C₅H₃ClIN
Peso molecolare: 239,4415 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD08235133
CAS Number: 77332-90-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR