Stampa…

(S)-2-Methylpyrrolidine hydrochloride

Fornitore: Apollo Scientific
Attenzione

OR460046-1G OR460046-25G OR460046-5G
APOSOR460046-1GEA 62.6 EUR
APOSOR460046-1G APOSOR460046-25G APOSOR460046-5G
(S)-2-Methylpyrrolidine hydrochloride
(S)-2-Methylpyrrolidine hydrochloride
Formula: C₅H₁₂ClN
Peso molecolare: 121,6100 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD06796611
CAS Number: 174500-74-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR