Stampa…
3-Hydroxypiperidine hydrochloride
Fornitore: Apollo Scientific
Synonyms: 3-Hydroxypiperidine Hydrochloride
OR460040-100G OR460040-25G OR460040-5G
APOSOR460040-100GEA 123 EUR
APOSOR460040-100G APOSOR460040-25G APOSOR460040-5G
3-Hydroxypiperidine hydrochloride
3-Hydroxypiperidine hydrochloride
Formula: C₅H₁₂ClNO
Peso molecolare: 137,61 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012772
CAS Number: 64051-79-2