Stampa…

3-Methylphenethyl bromide 97%

Fornitore: Apollo Scientific

OR452069-10G OR452069-1G OR452069-5G
APOSOR452069-10GEA 676 EUR
APOSOR452069-10G APOSOR452069-1G APOSOR452069-5G
3-Methylphenethyl bromide 97%
3-Methylphenethyl bromide
Formula: C₉H₁₁Br
Peso molecolare: 199.09 g/mol
Temperatura di stoccaggio: Ambiente
CAS Number: 16799-08-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR