Stampa…

Boc-D-Tyr(ME)-OH 97%

Fornitore: Apollo Scientific

Synonyms: Boc-D-Phe(4-OMe)-OH, N-Boc-4-methoxy-D-phenylalanine, N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine

OR452000-1G OR452000-25G OR452000-5G
APOSOR452000-1GEA 68.4 EUR
APOSOR452000-1G APOSOR452000-25G APOSOR452000-5G
Boc-D-Tyr(ME)-OH 97%
Boc-D-Tyr(ME)-OH
Formula: C₁₅H₂₁NO₅
Peso molecolare: 295,335 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00065604
CAS Number: 68856-96-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR