Stampa…

(S)-2-Amino-2-cyclopentylacetic acid 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: H-Cyclopentyl-Gly-OH, L-Cyclopentylglycine

OR451207-10G OR451207-5G OR451207-1G
APOSOR451207-10GEA 478 EUR
APOSOR451207-10G APOSOR451207-5G APOSOR451207-1G
(S)-2-Amino-2-cyclopentylacetic acid 98%
(S)-2-Amino-2-cyclopentylacetic acid
Formula: C₇H₁₃NO₂
Peso molecolare: 143,18 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03788076
CAS Number: 2521-84-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR