Stampa…

tert-Butyl (1-(hydroxymethyl)cyclobutyl)carbamate 95%

Fornitore: Apollo Scientific
Attenzione

Synonyms: N-Boc-1-(hydroxymethyl)cyclobutylamine

OR451134-1G OR451134-250MG
APOSOR451134-1GEA 271 EUR
APOSOR451134-1G APOSOR451134-250MG
tert-Butyl (1-(hydroxymethyl)cyclobutyl)carbamate 95%
tert-Butyl (1-(hydroxymethyl)cyclobutyl)carbamate
Formula: C₁₀H₁₉NO₃
Peso molecolare: 201,2628 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12028429
CAS Number: 1142211-17-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR