3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulphonyl chloride
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
3-Methyl-2-oxo-2,3-dihydrobenzo[d]oxazole-6-sulphonyl chloride
OR45030-1G
OR45030-250MG
APOSOR45030-1GEA
434
EUR
APOSOR45030-1G
APOSOR45030-250MG
3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulphonyl chloride
3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulphonyl chloride
Formula:
C₈H₆ClNO₄S Peso molecolare: 247,66 g/mol Punto di fusione: 137…139 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03033702 CAS Number: 62522-63-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...