Stampa…
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 90%
Fornitore: Apollo Scientific
Attenzione
Synonyms: Benzyl(methoxymethyl)[(trimethylsilyl)methyl]amine
OR43654-100G OR43654-25G OR43654-500G
APOSOR43654-100GEA 97.3 EUR
APOSOR43654-100G APOSOR43654-25G APOSOR43654-500G
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 90%
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine
Formula: C₁₃H₂₃NOSi
Punto di ebollizione: 76 °C/0,33mmHg
Densità: 0,928 g/cm³ (25 °C)
Flash Pt: 66 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00674005
CAS Number: 93102-05-7