1-Cyclopropyl-4-piperidinone 95+%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
1-Cyclopropylpiperidin-4-one
OR43592-10G
OR43592-5G
APOSOR43592-10GEA
108
EUR
APOSOR43592-10G
APOSOR43592-5G
1-Cyclopropyl-4-piperidinone 95+%
1-Cyclopropyl-4-piperidinone
Formula:
C₈H₁₃NO Peso molecolare: 139,20 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD07374375 CAS Number: 62813-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...