N-Methyldicyclohexylamine 97%
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
N,N-Dicyclohexylmethylamine, N-Cyclohexyl-N-methylcyclohexanamine
OR42279-100G
OR42279-500G
APOSOR42279-100GEA
17.6
EUR
APOSOR42279-100G
APOSOR42279-500G
N-Methyldicyclohexylamine 97%
N-Methyldicyclohexylamine
Formula:
C₁₃H₂₅N Peso molecolare: 195,3443 g/mol Punto di ebollizione: 265 °C Densità: 0,912 g/cm³ (25 °C) Flash Pt: 110 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00014289 CAS Number: 7560-83-0 UN: 2735 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...