Stampa…

2-Amino-6-chloronicotinic acid 97%

Fornitore: Apollo Scientific

OR42132-1G OR42132-5G
APOSOR42132-1GEA 74.2 EUR
APOSOR42132-1G APOSOR42132-5G
2-Amino-6-chloronicotinic acid 97%
2-Amino-6-chloronicotinic acid
Formula: C₆H₅ClN₂O₂
Peso molecolare: 172,5700 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11045328
CAS Number: 58584-92-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR