1-(4-Methoxybutyl)piperazine
Fornitore: Apollo Scientific
|
Attenzione
|
OR4157-1G
OR4157-2G
OR4157-5G
APOSOR4157-1GEA
52.4
EUR
APOSOR4157-1G
APOSOR4157-2G
APOSOR4157-5G
1-(4-Methoxybutyl)piperazine
1-(4-Methoxybutyl)piperazine
Formula:
C₉H₂₀N₂O Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02093551 CAS Number: 496808-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...