4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester 97%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(tert-Butoxycarbonyl)phenylboronic acid, pinacol ester, 4-Boc-phenylboronic acid Pinacol ester, 2-(4-tert-Butoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid tert-Butyl ester, 4-(tert-Butoxycarbonyl)phenylboronic acid Pinacol ester
Formula:
C₁₇H₂₅BO₄ Peso molecolare: 304,1890 g/mol Punto di fusione: 128…132 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03453055 CAS Number: 850568-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...