Stampa…

3-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride

Fornitore: Apollo Scientific
Pericolo

OR40700-1G OR40700-250MG
APOSOR40700-1GEA 357 EUR
APOSOR40700-1G APOSOR40700-250MG
3-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride
3-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride
Formula: C₅H₈Cl₂N₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD15143224
CAS Number: 88529-80-0
UN: 3261
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR