6-Bromo-4H-imidazo[4,5-b]pyridine 96%
Fornitore: Apollo Scientific
|
Attenzione
|
OR40185-1G
OR40185-5G
APOSOR40185-1GEA
96
EUR
APOSOR40185-1G
APOSOR40185-5G
6-Bromo-4H-imidazo[4,5-b]pyridine 96%
6-Bromo-4H-imidazo[4,5-b]pyridine
Formula:
C₆H₄BrN₃ Peso molecolare: 198,0200 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD08706033 CAS Number: 28279-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...