Stampa…

Methyl-2-bromo-5-methylthiazole-4-carboxylate

Fornitore: Apollo Scientific
Attenzione

Synonyms: Methyl 2-bromo-5-methylthiazole-4-carboxylate

OR40094-1G OR40094-250MG
APOSOR40094-1GEA 139 EUR
APOSOR40094-1G APOSOR40094-250MG
Methyl-2-bromo-5-methylthiazole-4-carboxylate
Methyl-2-bromo-5-methylthiazole-4-carboxylate
Formula: C₆H₆BrNO₂S
Peso molecolare: 236,09 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD14702517
CAS Number: 56355-61-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR